Cyanine 5 monosuccinimidyl ester ≥90% [equivalent to Cy5® NHS ester]
Lieferant: AAT BIOQUEST
Synonyme:
2-(5-(1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate, CY5-SE, Cy5 NHS ester
151
AATB151EA
152
EUR
AATB151
Cyanine 5 monosuccinimidyl ester ≥90% [equivalent to Cy5® NHS ester]
Cyanine 5 monosuccinimidyl ester
A variety of cyanine 5 ( Cy5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Multiple Bioconjugation Applications
- Excellent Quality
Bitte beachten For research use only.
Formel:
C₃₇H₄₃N₃O₁₀S₂ Molecular Weight: 753,8814 g/mol |
MDL:
MFCD29472578 CAS-Nummer: 146368-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...