Cyanine 5 maleimide ≥90% [equivalent to Cy5® maleimide]
Lieferant: AAT BIOQUEST
Synonyme: Cy5 maleimide
152
AATB152EA 307 EUR
AATB152
Cyanine 5 maleimide ≥90% [equivalent to Cy5® maleimide]
Cyanine 5 maleimide
A variety of cyanine 5 ( Cy5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Bitte beachten For research use only.