Dihydroethidium ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine

15200
AATB15200EA 152 EUR
AATB15200
Dihydroethidium ≥95%
Dihydroethidium

Hydroethidine operates effectively as a probe for measurement of reactive oxygen species.

Formel: C₂₁H₂₁N₃
Molecular Weight: 315,412 g/mol
MDL: MFCD00077335
CAS-Nummer: 104821-25-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR