2',7'-Dichlorofluorescin diacetate ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
H2DCFDA, 2,7-Dichlorodihydrofluorescein diacetate, DCDHF diacetate
15204
AATB15204EA
120
EUR
AATB15204
2',7'-Dichlorofluorescin diacetate ≥95%
2',7'-Dichlorofluorescin diacetate
DCFH-DA, 2',7'-Dichlorodihydrofluorescein diacetate (also called 2',7'-dichlorofluorescin diacetate), is hydrolyzed by cellular esterases to 2',7'-dichlorodihydrofluorescein (also called 2',7'-dichlorofluorescin) and is then oxidized to 2',7'-dichlorofluorescein primarily by H2O2.
Formel:
C₂₄H₁₆Cl₂O₇ Molecular Weight: 487,285 g/mol |
MDL:
MFCD00128955 CAS-Nummer: 4091-99-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...