1,2-Diamino-9,10-anthrachinon ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
1,2-Diaminoanthraquinone, 1,2-Diamino-10-anthracenedione
15220
AATB15220EA
152
EUR
AATB15220
1,2-Diamino-9,10-anthrachinon ≥95%
1,2-Diamino-9,10-anthrachinon
The role of nitric oxide (NO) as a putative mediator of neuronal death can be understood best if NO is detected directly.
Formel:
C₁₄H₁₀N₂O₂ Molecular Weight: 238,241 g/mol |
MDL:
MFCD00001219 CAS-Nummer: 1758-68-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...