2,3-Diaminonaphthalin ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Naphthalin-2,3-diamin, 2,3-Naphthylendiamin

15221
AATB15221EA 120 EUR
AATB15221
2,3-Diaminonaphthalin ≥95%
2,3-Diaminonaphthalin

The role of nitric oxide (NO) as a putative mediator of neuronal death can be understood best if NO is detected directly.

Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
MDL: MFCD00004116
CAS-Nummer: 771-97-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR