MCLA

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-(4-Methoxyphenyl)-2-methyl-3,7-dihydroimidazo[1,2-a]pyrazin-3-one Hydrochloride, 2-Methyl-6-(4-methoxyphenyl)-3,7-dihydroimidazo[1,2-a]pyrazin-3-one Hydrochloride

15250
AATB15250EA 152 EUR
AATB15250
MCLA
MCLA

MCLA has been used to assess superoxide formation in a variety of conditions, including neutrophil and macrophage activation, cultured cells and vascular tissue.

Formel: C₁₄H₁₃N₃O₂
Molecular Weight: 291,74 g/mol
MDL: MFCD00144728
CAS-Nummer: 128322-44-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR