ReadiUse™ NADP Regenerating Kit
Lieferant: AAT BIOQUEST
15266
AATB15266EA
387
EUR
AATB15266
ReadiUse™ NADP Regenerating Kit
Tests
Enzymtests
Designed for consistent and dependable results. AAT Bioquest's ReadiUse™ NADP regenerating kit provides a flexible prepackaged NADP regenerating system to accommodate various experimental designs.
- Uses two ready-to-use solutions to regenerate NADP by a simple mixing
- 300-500 enzyme assays can be performed using this kit
- Six-month shelf life
This kit can be used for all NADP-requiring systems (such as: 12-Ketochenodeoxycholic Acid synthesis, Ferredoxin Reductase System).
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.