Resazurin Natriumsalz ≥90%

Lieferant: AAT BIOQUEST
Achtung

15700
AATB15700EA 120 EUR
AATB15700
Resazurin Natriumsalz ≥90%
Resazurin Natriumsalz

Resazurin is a non-toxic, water-soluble and redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by many biological processes.

Formel: C₁₂H₆NNaO₄
MDL: MFCD00005036
CAS-Nummer: 62758-13-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR