MitoROS Brite™ 570 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
Lieferant: AAT BIOQUEST
15998
AATB15998EA
334
EUR
AATB15998
MitoROS Brite™ 570 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
MitoROS Brite™ 570
The cell-permeant MitoROS Brite™ 570 reagent is cell-permeable and selectively located in mitochondria.
- Stable for 12 months
Mitochondrial ROS (mtROS or mROS) are reactive oxygen species (ROS) produced in mitochondria.
Bitte beachten For Research Use Only