MitoROS Brite™ 670 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
Lieferant: AAT BIOQUEST
15999
AATB15999EA
334
EUR
AATB15999
MitoROS Brite™ 670 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
MitoROS Brite™ 670
The cell-permeant MitoROS Brite™ 670 reagent is cell-permeable and selectively located in mitochondria.
- Stable for 12 months
Mitochondrial ROS (mtROS or mROS) are reactive oxygen species (ROS) produced in mitochondria.
Bitte beachten For Research Use Only