Cyanine 7 monoacid ≥90% [equivalent to Cy7® acid]

Lieferant: AAT BIOQUEST

Synonyme: Cy7 acid

160
AATB160EA 383 EUR
AATB160
Cyanine 7 monoacid ≥90% [equivalent to Cy7® acid]
Cyanine 7 monoacid

A variety of cyanine 7 (Cy7®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • Multiple Bioconjugation Applications
  • Excellent Quality


Bitte beachten For research use only.

CAS-Nummer: 943298-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR