ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
Lieferant: AAT BIOQUEST
|
Achtung
|
16002
AATB16002EA
307
EUR
AATB16002
ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
ROS Brite™
Reactive oxygen species (ROS) are chemically reactive molecules containing oxygen.
- Bright fluorescence
- Useful ros probe
- Multiple research applications
Bitte beachten For research use only.
Molecular Weight:
732,81 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...