ROS Brite™ 700 Optimized for in Vivo Imaging

Lieferant: AAT BIOQUEST
Achtung

16004
AATB16004EA 307 EUR
AATB16004
ROS Brite™ 700 Optimized for in Vivo Imaging
ROS Brite™

Reactive oxygen species (ROS) are chemically reactive molecules containing oxygen.


  • Water soluble
  • Cell impermeant
  • Bright nir signal
  • Useful for in vivo imaging


Bitte beachten For research use only.

Molecular Weight: 732,81 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR