MitoROS Brite™ NIR 780 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
Lieferant: AAT BIOQUEST
16049
AATB16049EA 402 EUR
AATB16049
MitoROS Brite™ NIR 780 [Optimized for Detecting Reactive Oxygen Species (ROS) in Mitochondria]
MitoROS Brite™ NIR 780
MitoROS Brite™ NIR 780 is a superoxide-sensitive fluorescent probe specifically designed for targeting mitochondria in live cells.
  • Stable for 12 months
Mitochondrial reactive oxygen species (mtROS or mROS) are generated within mitochondria, primarily during oxidative phosphorylation at the electron transport chain (ETC) on the inner mitochondrial membrane.
Bitte beachten For Research Use Only