MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Lieferant: AAT BIOQUEST
16052
AATB16052EA 307 EUR
AATB16052
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Proteinkonjugate und Markierungen
Oxidation of MitoROS™ 580 by superoxide generates red fluorescence.
  • Optimized for mitochondria
  • Live cell reagent
MitoROS™ 580 reagent permeates live cells where it selectively targets mitochondria. It is rapidly oxidized by superoxide. It is less likely to be oxidized by other reactive oxygen species (ROS) and reactive nitrogen species (RNS). The oxidized product is highly fluorescent in cells.
Verpackung: Plastic vial.
Bitte beachten For research use only.