ROS Brite™ DHCF

Lieferant: AAT BIOQUEST
Achtung

16053
AATB16053EA 307 EUR
AATB16053
ROS Brite™ DHCF
ROS Brite™

Reactive oxygen species (ROS) are chemically reactive molecules containing oxygen.


  • Bright fluorescence
  • Useful ros probe
  • Multiple research applications


Bitte beachten For research use only.

Molecular Weight: 732,81 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR