Cyanine 7 monosuccinimidyl ester ≥90% [equivalent to Cy7® NHS ester]
Lieferant: AAT BIOQUEST
Synonyme:
Cy7 NHS ester, 2-(7-(1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)hepta-1,3,5-trien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate, CY7-SE
161
AATB161EA
152
EUR
AATB161
Cyanine 7 monosuccinimidyl ester ≥90% [equivalent to Cy7® NHS ester]
Cyanine 7 monosuccinimidyl ester
A variety of cyanine 7 (Cy7®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Multiple Bioconjugation Applications
- Excellent Quality
Bitte beachten For research use only.
Formel:
C₃₉H₄₅N₃O₁₀S₂ Molecular Weight: 779,92 g/mol |
MDL:
MFCD28968779 CAS-Nummer: 477908-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...