mFluor™ Violet 530 SE
Lieferant: AAT BIOQUEST
1616
AATB1616EA
309
EUR
AATB1616
mFluor™ Violet 530 SE
mFluor™ Violet 530 SE
mFluor™ Violet 530 dyes have fluorescence excitation and emission maxima of ~405 nm and ~530 nm respectively.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only