mFluor™ Violet 530 maleimide
Lieferant: AAT BIOQUEST
1617
AATB1617EA
420
EUR
AATB1617
mFluor™ Violet 530 maleimide
mFluor™ Violet 530 maleimide
mFluor™ Violet 530 dyes have fluorescence excitation and emission maxima of ~405 nm and ~530 nm respectively.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only