Cyanine 7 maleimide ≥90% [equivalent to Cy7® maleimide]
Lieferant: AAT BIOQUEST
Synonyme:
Cy7 maleimide
162
AATB162EA
383
EUR
AATB162
Cyanine 7 maleimide ≥90% [equivalent to Cy7® maleimide]
Cyanine 7 maleimide
A variety of cyanine 7 (Cy7®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Multiple Bioconjugation Applications
- Excellent Quality
Bitte beachten For research use only.