DAF-2 zur Stickoxiderkennung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2-​(3,​6-​dihydroxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, 4,5-Diaminofluorescein

16290
AATB16290EA 439 EUR
AATB16290
DAF-2 zur Stickoxiderkennung
DAF-2

The weakly fluorescent DAF-2 is a sensitive fluorogenic indicator commonly used for detecting nitric oxide (NO).


  • Sensitive NO detection

Formel: C₂₀H₁₆N₂O₅
Molecular Weight: 364,34 g/mol
MDL: MFCD01866182
CAS-Nummer: 205391-01-1
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd4fd7fff95bd52e7c1e414856d02702d141dea5&vid=6e26b1c7f984870793c18e075c2ee6c3e4908c0c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR