Cyanine 7 azide ≥95% [equivalent to Cy7® azide]

Lieferant: AAT BIOQUEST

Synonyme: Cy7 azide

163
AATB163EA 383 EUR
AATB163
Cyanine 7 azide ≥95% [equivalent to Cy7® azide]
Cyanine 7 azide

A variety of cyanine 7 (Cy7®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • Multiple Bioconjugation Applications
  • Excellent Quality


Bitte beachten For research use only.

Molecular Weight: 864,95 g/mol
CAS-Nummer: 1557149-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR