Amplite® Fluorimetric Peroxynitrite Quantification Kit *Green Fluorescence*
Lieferant: AAT BIOQUEST
16316
AATB16316EA
537
EUR
AATB16316
Amplite® Fluorimetric Peroxynitrite Quantification Kit *Green Fluorescence*
Tests
Enzymtests
Peroxynitrite (ONOO-) is a strong oxidizing species and a highly active nitrating agent.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.