Amplite® Fluorimetric Peroxynitrite Quantification Kit *Green Fluorescence*
Lieferant: AAT BIOQUEST
16316
AATB16316EA 537 EUR
AATB16316
Amplite® Fluorimetric Peroxynitrite Quantification Kit *Green Fluorescence*
Tests Enzymtests
Peroxynitrite (ONOO-) is a strong oxidizing species and a highly active nitrating agent.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.