mFluor™ UV420 SE
Lieferant: AAT BIOQUEST
1641
AATB1641EA
429
EUR
AATB1641
mFluor™ UV420 SE
mFluor™ UV420 SE
mFluor™ UV 420 dyes have fluorescence excitation and emission maxima of ~350 nm and ~420 nm respectively.
- Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only