mFluor™ UV520 SE
Lieferant: AAT BIOQUEST
1643
AATB1643EA
429
EUR
AATB1643
mFluor™ UV520 SE
mFluor™ UV520 SE
mFluor™ UV 520 dyes have fluorescence excitation and emission maxima of ~350 nm and ~520 nm respectively.
- Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only