mFluor™ UV540 SE
Lieferant: AAT BIOQUEST
1645
AATB1645EA 429 EUR
AATB1645
mFluor™ UV540 SE
mFluor™ UV540 SE
mFluor™ UV 540 dyes have fluorescence excitation and emission maxima of ~350 nm and ~540 nm respectively.
  • Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only