mFluor™ UV610 SE
Lieferant: AAT BIOQUEST
1649
AATB1649EA 429 EUR
AATB1649
mFluor™ UV610 SE
mFluor™ UV610 SE
mFluor™ UV 610 dyes have fluorescence excitation and emission maxima of ~350 nm and ~610 nm respectively.
  • Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Bitte beachten For Research Use Only