Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Lieferant: AAT BIOQUEST
167
AATB167EA 387 EUR
AATB167
Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Cyanine 7.5 monoacid
Cy7.5 dyes undergo comprehensive quality control testing to ensure high levels of chromophore and reactive dye content.
  • Stable for 24 months
Various cyanine dyes have been extensively used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analyses.
Bitte beachten For Research Use Only