Cyanine 7.5 monosuccinimidyl ester [equivalent to Cy7.5® NHS ester]
Lieferant: AAT BIOQUEST
Synonyme:
Cy7.5 NHS ester
168
AATB168EA
153
EUR
AATB168
Cyanine 7.5 monosuccinimidyl ester [equivalent to Cy7.5® NHS ester]
Cyanine 7.5 monosuccinimidyl ester
Cy7.5 dyes undergo comprehensive quality control testing to ensure high levels of chromophore and reactive dye content.
- Stable for 12 months
Various cyanine dyes have been extensively used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analyses.
Bitte beachten For Research Use Only