Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 350
Lieferant: AAT BIOQUEST
16890
AATB16890EA
230
EUR
AATB16890
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 350
Proteine und Peptide
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
- Extremely high biotin-binding affinity
- High quality conjugate
Bitte beachten For research use only.