Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 594
Lieferant: AAT BIOQUEST
16892
AATB16892EA 230 EUR
AATB16892
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 594
Proteine und Peptide
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Extremely high biotin-binding affinity
  • High quality conjugate
Bitte beachten For research use only.