Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
Lieferant: AAT BIOQUEST
16906
AATB16906EA
292
EUR
AATB16906
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
Proteine und Peptide
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Extremely high biotin-binding affinity
- High quality conjugate
Bitte beachten For research use only.