Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 510
Lieferant: AAT BIOQUEST
16931
AATB16931EA 152 EUR
AATB16931
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 510
Proteine und Peptide
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Optimized for flow cytometry
  • Useful for assay development
  • High affinity
  • Multiple applications
Bitte beachten For research use only.