Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Blue 570
Lieferant: AAT BIOQUEST
16935
AATB16935EA
152
EUR
AATB16935
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Blue 570
Proteine und Peptide
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Optimized for flow cytometry
- Useful for assay development
- High affinity
- Multiple applications
Bitte beachten For research use only.