Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 790
Lieferant: AAT BIOQUEST
16975
AATB16975EA 152 EUR
AATB16975
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 790
Proteine und Peptide
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
  • Extremely high biotin-binding affinity
  • High quality conjugate
Bitte beachten For research use only.