Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800
Lieferant: AAT BIOQUEST
16978
AATB16978EA
152
EUR
AATB16978
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800
Proteine und Peptide
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Extremely high biotin-binding affinity
- High quality conjugate
Bitte beachten For research use only.