XFD488 alkyne fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1700
AATB1700EA 460 EUR
AATB1700
XFD488 alkyne fluoreszierende Probe
XFD488 alkyne

The fluorescent dye alkynes react with azides via a copper-catalyzed click reaction (CuAAC).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 773,92 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR