TF2-dUTP 1 mM in TRIS Puffer (pH 7.5)
Lieferant: AAT BIOQUEST
|
Achtung
|
17007
AATB17007EA
460
EUR
AATB17007
TF2-dUTP 1 mM in TRIS Puffer (pH 7.5)
TF2-dUTP
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- Useful for fish probe development
- Well-cited
- Multiple applications
Bitte beachten For research use only.
Molecular Weight:
1100 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...