TF3-dUTP 1 mM in TRIS Puffer (pH 7.5)

Lieferant: AAT BIOQUEST
Achtung

17008
AATB17008EA 460 EUR
AATB17008
TF3-dUTP 1 mM in TRIS Puffer (pH 7.5)
TF3-dUTP

The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • Useful for fish probe development
  • Well-cited
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 1150 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR