XFD488 azide fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1701
AATB1701EA 460 EUR
AATB1701
XFD488 azide fluoreszierende Probe
XFD488 azide

The fluorescent dye azides react with alkynes via a copper-catalyzed click reaction (CuAAC).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 861,04 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR