Violet 450-dUTP 1 mM in TRIS Puffer, mFluor™ (pH 7.5)
Lieferant: AAT BIOQUEST
Achtung
17011
AATB17011EA 460 EUR
AATB17011
Violet 450-dUTP 1 mM in TRIS Puffer, mFluor™ (pH 7.5)
Violet 450-dUTP
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
  • Useful for fish probe development
  • Well-cited
  • Multiple applications
Bitte beachten For research use only.
ADR: 5,II