Digoxigenin-11-dUTP 1 mM für PCR

Lieferant: AAT BIOQUEST
Achtung

17012
AATB17012EA 230 EUR
AATB17012
Digoxigenin-11-dUTP 1 mM für PCR
Digoxigenin-11-dUTP

Digoxigenin-11-dUTP can be enzymatically incorporated into DNA/cDNA as substitute for its natural counterpart dTTP.


  • Limited DIG interference
  • High quality

Molecular Weight: 1132,87 g/mol
CAS-Nummer: 129030-36-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df1f0cdc5162010d478efc4a2fdfb2a4e1c3c3d5&vid=714a02683f179f9416f60adc4688369dd1e562cf&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR