Biotin-11-dGTP 1 mM in TRIS Puffer (pH 7.5)
Lieferant: AAT BIOQUEST
|
Achtung
|
17015
AATB17015EA
143
EUR
AATB17015
Biotin-11-dGTP 1 mM in TRIS Puffer (pH 7.5)
Biotin-11-dGTP
The biotin-modified deoxyguanosine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.
- Useful for DNA probe development
- Well-cited
- Multiple applications
Bitte beachten For research use only.
Molecular Weight:
898,71 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...