Biotin-11-dUTP 1 mM in TRIS Puffer ≥95% (pH 7.5)

Lieferant: AAT BIOQUEST
Achtung

17016
AATB17016EA 152 EUR
AATB17016
Biotin-11-dUTP 1 mM in TRIS Puffer ≥95% (pH 7.5)
Biotin-11-dUTP

The biotin-modified deoxyuridine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.

Formel: C₂₈H₄₅N₆O₁₇P₃S
CAS-Nummer: 86303-25-5
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR