Biotin-16-dUTP 1 mM in TRIS Puffer ≥95% pH 7.5
Lieferant: AAT BIOQUEST
Achtung
17017
AATB17017EA 152 EUR
AATB17017
Biotin-16-dUTP 1 mM in TRIS Puffer ≥95% pH 7.5
Biotin-16-dUTP
The biotin-modified deoxyuridine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.
CAS-Nummer: 136632-31-0
ADR: 5,II