Biotin-20-dUTP 1 mM in TRIS Puffer (pH 7.5)
Lieferant: AAT BIOQUEST
|
Achtung
|
17018
AATB17018EA
152
EUR
AATB17018
Biotin-20-dUTP 1 mM in TRIS Puffer (pH 7.5)
Biotin-20-dUTP
The biotin-modified deoxyuridine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.
- Useful for DNA probe development
- Well-cited
- Multiple applications
Bitte beachten For research use only.
ADR:
5,II |