Biotin-14-dCTP 1 mM in TRIS Puffer (pH 7.5)

Lieferant: AAT BIOQUEST
Achtung

17019
AATB17019EA 152 EUR
AATB17019
Biotin-14-dCTP 1 mM in TRIS Puffer (pH 7.5)
Biotin-14-dCTP

The biotin-modified dCTP analogs are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 905,79 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR