Fluorescein-12-dUTP sodium salt 1 mM in TRIS Puffer ≥95% pH 7.5
Lieferant: AAT BIOQUEST
|
Achtung
|
17022
AATB17022EA
152
EUR
AATB17022
Fluorescein-12-dUTP sodium salt 1 mM in TRIS Puffer ≥95% pH 7.5
Fluorescein-12-dUTP sodium salt
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
Formel:
C₃₉H₃₈N₄O₂₁P₃Na₃ Molecular Weight: 1060,62 g/mol |
CAS-Nummer:
214154-36-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...