Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS Puffer (pH 7.5)
Lieferant: AAT BIOQUEST
Achtung
17025
AATB17025EA 143 EUR
AATB17025
Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS Puffer (pH 7.5)
Cyanine-3- dUTP [Cy3-dUTP]
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
  • Useful for DNA probe development
  • Well-cited
  • Multiple applications
Bitte beachten For research use only.
ADR: 5,II